Learn More
Apexbio Technology LLC Cy7.5 azide (non-sulfonated), 5mg. Cas: MFCD: NA. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A81165
Encompass_Preferred
Cy7.5 azide a NIR fluorescence dye that ready for the use in Click Chemistry labeling with long-wave infrared fluorescence. For other fluorescent applications, especially requiring low fluorescent background, this labeling reagent was also popular used. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5017