Learn More
Medchemexpress LLC Cy7.5-COOH bromide | 2978106-25-9 | 95.2% | 10 MG
SDP
Catalog No. 5000297088
Encompass_Preferred
Cy7.5-COOH (bromide) is a near-infrared cyanine fluorescent dye supplied as the bromide salt. It exhibits absorbance/emission maxima near 781/808 nm and is used as a fluorescent probe in DNA sequencing, flow cytometry, fluorescence microscopy, and in vivo imaging.
  • High near-infrared fluorescence (Abs/Em = 781/808 nm).
  • Suitable as a fluorescent probe for DNA sequencing, flow cytometry, and in vivo imaging.
  • Provided as the bromide salt to support solubility in common organic solvents.
  • Available in small research quantities for laboratory assays and labeling.
  • Compatible with standard conjugation and labeling workflows.
Catalog No. 50-002-97088 Supplier Medchemexpress LLC Supplier No. HYD1883B10MG
Cy7.5-COOH (bromide) is a near-infrared cyanine fluorescent dye supplied as the bromide salt. It exhibits absorbance/emission maxima near 781/808 nm and is used as a fluorescent probe in DNA sequencing, flow cytometry, fluorescence microscopy, and in vivo imaging.
  • High near-infrared fluorescence (Abs/Em = 781/808 nm).
  • Suitable as a fluorescent probe for DNA sequencing, flow cytometry, and in vivo imaging.
  • Provided as the bromide salt to support solubility in common organic solvents.
  • Available in small research quantities for laboratory assays and labeling.
  • Compatible with standard conjugation and labeling workflows.