Learn More
Apexbio Technology LLC Cy7 azide (non-sulfonated), 25mg. Cas: MFCD: NA. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A811525
Encompass_Preferred
Cy7 azide is a labeling reagent that ready for the use in Click Chemistry labeling. This labeling dye is a near-infrared dye which has low aqueous solubility. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5014