Learn More
Apexbio Technology LLC Cy7 carboxylic acid (non-sulfonated), 25mg
SDP
Catalog No. 501905046
Encompass_Preferred
Cy7 carboxylic acid is a near-infrared fluorescent dye which has low aqueous solubility. As for coupling and labeling reactions, pre-activated Cy7 NHS ester or water-soluble sulfo-Cy7 NHS ester would be considered. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5046 Supplier Apexbio Technology LLC Supplier No. A813525
Cy7 carboxylic acid is a near-infrared fluorescent dye which has low aqueous solubility. As for coupling and labeling reactions, pre-activated Cy7 NHS ester or water-soluble sulfo-Cy7 NHS ester would be considered. Other sizes are available. Please inqury us for quote.