Learn More
Medchemexpress LLC HY-D1048 5mg , Cy7 DiC18 CAS:100068-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD10485MG
Encompass_Preferred
HY-D1048 5mg Cy7 DiC18 CAS: 100068-60-8 Cy7 DiC18 is a lipophilic, near-infrared fluorescent cyanine dye used for staining membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1938