Learn More
Medchemexpress LLC Cy7 hydrazide | 2183440-63-1 | MFCD31580092 | 90.0% | 10 MG
SDP
Catalog No. 5000297691
Encompass_Preferred
Cy7 hydrazide is a cyanine-derived near-infrared fluorescent hydrazide used to form hydrazone linkages with aldehyde- or ketone-containing molecules for fluorescence labeling and imaging applications. It is supplied as a brown to black solid and should be stored sealed, away from moisture and light.
Catalog No. 50-002-97691 Supplier Medchemexpress LLC Supplier No. HYD250210MG
Cy7 hydrazide is a cyanine-derived near-infrared fluorescent hydrazide used to form hydrazone linkages with aldehyde- or ketone-containing molecules for fluorescence labeling and imaging applications. It is supplied as a brown to black solid and should be stored sealed, away from moisture and light.