Learn More
Medchemexpress LLC CY7-N3 (Sulfo-Cyanine7-N3) | 97.6% | 763.95 | 5 MG
SDP
Catalog No. 5000429139
Encompass_Preferred
CY7-N3 (Sulfo-Cyanine7-N3) is a water-soluble Near-Infrared (NIR) dye azide designed for Click Chemistry. It possesses an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Water-soluble NIR dye azide
  • Contains an azide group for Click Chemistry
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-004-29139 Supplier Medchemexpress LLC Supplier No. HYD10535MG
CY7-N3 (Sulfo-Cyanine7-N3) is a water-soluble Near-Infrared (NIR) dye azide designed for Click Chemistry. It possesses an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Water-soluble NIR dye azide
  • Contains an azide group for Click Chemistry
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC)