Learn More
Lumiprobe Cyanine7 NHS ester, 1432466-81-3, 1mg
Catalog No. NC0201529
Encompass_Preferred
Molecular formula: C41H48N3BF4O4, Molecular weight: 733.64, Appearance: dark green solid, Solubility: soluble in organic solvents (DMSO, DMF, dichloromethane), low solubility in water; Cyanine7 is a near infrared dye for bioimaging and sonoluminiscence applications. The dye exhibits high extinction coefficient to provide high contrast images. It can be conjugated to amine groups of proteins, peptides, and small molecules.
Catalog No. NC0201529 Supplier Lumiprobe Supplier No. 15020
Molecular formula: C41H48N3BF4O4, Molecular weight: 733.64, Appearance: dark green solid, Solubility: soluble in organic solvents (DMSO, DMF, dichloromethane), low solubility in water; Cyanine7 is a near infrared dye for bioimaging and sonoluminiscence applications. The dye exhibits high extinction coefficient to provide high contrast images. It can be conjugated to amine groups of proteins, peptides, and small molecules.