Learn More
Apexbio Technology LLC Cy7 NHS ester, 25mg. Cas: MFCD: MFCD28385523. Labeling of amino-groups in biomolecules.
SDP
Supplier:  Apexbio Technology LLC A810925
Encompass_Preferred
Sulfo-Cy7 NHS ester is a sulfonated, hydrophilic and highly water soluble dye. This reagent allows prepare Cy7-labeled biomolecules, especially useful for the labeling of delicate proteins, and proteins prone to denaturation. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5002