Learn More
Apexbio Technology LLC Cy7 NHS ester (non-sulfonated), 25mg. Cas: 1432019-64-1 MFCD: NA. Labeling of amino-groups in biomolecules.
SDP
Supplier:  Apexbio Technology LLC A810425
Encompass_Preferred
Cy7 NHS ester, also called Cy7 monofunctional N-hydroxysuccinimide ester or Cy7-NHS [1], is a near-infrared fluorescent [2] and soluble sulfonated cyanine dye [3] with a molecular weight of 828 Da[2], able to be dissolved in DMSO [1] and ready for conjugation to proteins [4]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4992