Learn More
TARGETMOL CHEMICALS INC CY7-SE TRIETHYLAMINE 477 10MG
SDP
Catalog No. 502581063
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. CY7-SE Triethylamine is a fluorescence labeling agent (Ex=700-770 nm,Em=790 nm) used for labeling proteins peptides antibodies and oligonucleotides.
Catalog No. 50-258-1063 Supplier TARGETMOL CHEMICALS INC Supplier No. T18954L10MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. CY7-SE Triethylamine is a fluorescence labeling agent (Ex=700-770 nm,Em=790 nm) used for labeling proteins peptides antibodies and oligonucleotides.