Learn More
Medchemexpress LLC CY7-SE triethylamine (Sulfo-Cyanine7 Succinimidyl Ester triethylamine) | ≥98.0% | 5 MG
SDP
Catalog No. 5000409716
Encompass_Preferred
CY7-SE triethylamine is a CY dye, a type of Cyanine compound characterized by two nitrogen atoms linked by an odd number of methyl units. These compounds are known for their long wavelength, adjustable absorption and emission, high extinction coefficient, good water solubility, and relatively simple synthesis. CY dyes are frequently utilized for labeling proteins, antibodies, and small molecular compounds. The document provides information on the simple mixing reaction method for labeling protein antibodies, offering relevant guidance.
  • Long wavelength
  • Adjustable absorption and emission
  • High extinction coefficient
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling proteins, antibodies, and small molecular compounds
Catalog No. 50-004-09716 Supplier Medchemexpress LLC Supplier No. HYD0824A5MG
CY7-SE triethylamine is a CY dye, a type of Cyanine compound characterized by two nitrogen atoms linked by an odd number of methyl units. These compounds are known for their long wavelength, adjustable absorption and emission, high extinction coefficient, good water solubility, and relatively simple synthesis. CY dyes are frequently utilized for labeling proteins, antibodies, and small molecular compounds. The document provides information on the simple mixing reaction method for labeling protein antibodies, offering relevant guidance.
  • Long wavelength
  • Adjustable absorption and emission
  • High extinction coefficient
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling proteins, antibodies, and small molecular compounds