Learn More
Apexbio Technology LLC CY7-TRIETHYLAMMONIUM-SALT-1MG
SDP
Catalog No. 5000567878
Encompass_Preferred
Cy7 triethylammonium salt is a fluorescent dye commonly used in biomedical research particularly in imaging and diagnostic applications Originating from cyanine-based dye technology it operates by targeting cellular and molecular components enabling visualization in complex biological systems Its fluorescence mechanism involves energy absorption and emission within the near-infrared range allowing for deep tissue penetration with minimal background interference In vitro studies often reflect its activity in the nanomolar to low micromolar range making it suitable for high-sensitivity assays Researchers typically utilize this compound in various cell models and animal studies serving as an important tool for drug screening and tracking biological processes Experimental concentrations typically depend on the specific research design ensuring optimal performance and result accuracy
Catalog No. 50-005-67878 Supplier Apexbio Technology LLC Supplier No. C85921
Cy7 triethylammonium salt is a fluorescent dye commonly used in biomedical research particularly in imaging and diagnostic applications Originating from cyanine-based dye technology it operates by targeting cellular and molecular components enabling visualization in complex biological systems Its fluorescence mechanism involves energy absorption and emission within the near-infrared range allowing for deep tissue penetration with minimal background interference In vitro studies often reflect its activity in the nanomolar to low micromolar range making it suitable for high-sensitivity assays Researchers typically utilize this compound in various cell models and animal studies serving as an important tool for drug screening and tracking biological processes Experimental concentrations typically depend on the specific research design ensuring optimal performance and result accuracy