Learn More
Medchemexpress LLC Cy7-yne | 95.0% | 10 MG
SDP
Catalog No. 5000296975
Encompass_Preferred
Cy7-YNE is a near-infrared cyanine fluorescent labeling reagent containing an alkyne functional group for click chemistry and conjugation to biomolecules. It is used to label proteins, antibodies, and peptides for fluorescence-based detection in the ~700-790 nm spectral region.
  • Near-infrared cyanine dye suitable for biomolecule labeling.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Optimized for excitation around 700-770 nm and emission near 770-790 nm.
  • Available in milligram pack sizes for research use.
  • Solid form with ≥95.0% purity.
  • Store protected from light; in solvent: -80°C for long-term, -20°C for short-term.
Catalog No. 50-002-96975 Supplier Medchemexpress LLC Supplier No. HYD105210MG
Cy7-YNE is a near-infrared cyanine fluorescent labeling reagent containing an alkyne functional group for click chemistry and conjugation to biomolecules. It is used to label proteins, antibodies, and peptides for fluorescence-based detection in the ~700-790 nm spectral region.
  • Near-infrared cyanine dye suitable for biomolecule labeling.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Optimized for excitation around 700-770 nm and emission near 770-790 nm.
  • Available in milligram pack sizes for research use.
  • Solid form with ≥95.0% purity.
  • Store protected from light; in solvent: -80°C for long-term, -20°C for short-term.