Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanamide, 50% w/w aq. soln., stab.

Catalog No. aal2044822
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2044822 100 g
AAL2044830 250 g
AAL204480B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2044822 Supplier Thermo Scientific Chemicals Supplier No. L2044822
Only null left
Add to Cart

CAS: 420-04-2 | CH2N2 | 42.041 g/mol

It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.

Solubility
Soluble in water (85 g/100 ml (25°C). Soluble in organic solvents.

Notes
May form precipitate on storage
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420-04-2
Molecular Formula CH2N2
Molecular Weight (g/mol) 42.041
MDL Number MFCD00007572
InChI Key XZMCDFZZKTWFGF-UHFFFAOYSA-N
Synonym hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
PubChem CID 9864
ChEBI CHEBI:16698
IUPAC Name cyanamide
SMILES C(#N)N

Specifications

Density 1.06
pH 4.2
Flash Point None
Odor Odorless
Refractive Index 1.395
Linear Formula H2NCN
Quantity 100 g
UN Number UN2922
Beilstein 1732569
Sensitivity Light sensitive; May form a precipitate
Merck Index 14,2684
Solubility Information Soluble in water (85g/100 ml (25°C). Soluble in organic solvents.
Formula Weight 42.04
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Cyanamide, stabilized
Show More Show Less
Hazard Category H301+H311-H314-H317-H335-H351-H361fd-H373
Hazard Statement GHS H Statement
H301-H314-H312-H319-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 206-992-3
RTECSNumber GS5950000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.