Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanamide, 98+%, stab.

Catalog No. AAL0374422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0374422 100 g
AAL0374406 5 g
AAL0374414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0374422 Supplier Thermo Scientific Chemicals Supplier No. L0374422
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 420-04-2 | CH2N2 | 42.041 g/mol

Cyanamide is used as an intermediate in the production of pharmaceuticals and other organic compounds. It is also used as an alcohol deterrent drug. It is widely used as plant growth regulator, agrochemical and fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanamide is used as an intermediate in the production of pharmaceuticals and other organic compounds. It is also used as an alcohol deterrent drug. It is widely used as plant growth regulator, agrochemical and fertilizer.

Solubility
Soluble in water, butanol, methyl ethyl ketone, ethyl acetate, alcohols, phenols, chloroform, amines and ethers. Slightly soluble in carbon disufide. Insoluble in cyclohexane.

Notes
Incompatible with barium peroxide, boric acid, dry hydrogen and hydrogen peroxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420-04-2
Molecular Formula CH2N2
Molecular Weight (g/mol) 42.041
MDL Number MFCD00007572
InChI Key XZMCDFZZKTWFGF-UHFFFAOYSA-N
Synonym hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
PubChem CID 9864
ChEBI CHEBI:16698
IUPAC Name cyanamide
SMILES C(#N)N

Specifications

Melting Point 43°C to 47°C
Density 1.282
pH 3.5 to 4.2
Boiling Point 132°C to 138°C (12 mmHg)
Flash Point 141°C (285°F)
Odor Odorless
Linear Formula H2NCN
Quantity 100 g
UN Number UN2923
Beilstein 1732569
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,2684
Solubility Information Soluble in water,butanol,methyl ethyl ketone, ethyl acetate,alcohols,phenols,chloroform,amines and ethers. Slightly soluble in carbon disufide. Insoluble in cyclohexane.
Formula Weight 42.04
Percent Purity ≥98%
Chemical Name or Material Cyanamide, stabilized
Show More Show Less
Hazard Category H301+H311-H314-H317-H335-H351-H361fd-H373
Hazard Statement GHS H Statement
H301-H314-H312-H319-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 206-992-3
RTECSNumber GS5950000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.