Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanide ISE Fill Solution, Fisherbrand
SDP

Catalog No. ACCU0808500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0808500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0808500
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Cyanide ISE Fill Solution
CAS 7447-40-7, 7757-79-1, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Water Analysis
For Use With (Equipment) Ion Selective Electrode
Physical Form Liquid
Quantity 500 mL
Solution Type Ionic Strength Adjustor
Odor Odorless
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.