Learn More
Cyanide Standard, 0.1 ppm CN, Reagents
Specifications
Specifications
| Chemical Name or Material | Cyanide Standard, 0.1 ppm CN |
| Packaging | Opaque HDPE |
| CAS | 7732-18-5, 151-50-8 |
| Specific Gravity | 1.0001 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| For Use With (Equipment) | Cyanide Analyzer |
| Solubility | Completely miscible |
| Industry Type | Cyanide Analysis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.