Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanide Standard, 1000 ppm CN, Reagents

Catalog No. 01335698
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-698 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-698 Supplier Reagents Holdings Llc Supplier No. CC0060001B
Add to Cart
Add to Cart

Cyanide Standard, 1000 ppm CN, Reagent, use for Cyanide analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Cyanide Standard, 1000 ppm CN
Packaging Opaque HDPE
CAS 7732-18-5, 1310-73-2
Specific Gravity 1.022
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Cyanide Analyzer
Solubility Completely miscible
Industry Type Cyanide Analysis
Quantity 1 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.