Learn More
AccuStandard, Inc Cyanide Standard | 57-12-5 | MFCD00468042 | 100mL
SDP
Supplier:  AccuStandard, Inc WCCN1X1
Encompass
Cyanide Standard | 100 µg/mL in 2% Sodium hydroxide | MW: 26.018 | 57-12-5 | MFCD00468042 | 100mL
Catalog No. NC1826934