Learn More
Aqua Solutions Cyanide Standard (1% Solution) 10,000 ppm as HCN 1 ml = 10 mg HCN (250ML)
SDP
Catalog No. NC0120006
Encompass_Preferred
Cyanide Standard (1% Solution) 10,000 ppm as HCN 1 ml = 10 mg HCN (250ML)
Catalog No. NC0120006 Supplier Aqua Solutions Supplier No. DC915130
 Restricted Item
Add to Cart
Add to Cart
Cyanide Standard (1% Solution) 10,000 ppm as HCN 1 ml = 10 mg HCN (250ML)