Learn More
SPEX CERTIPREP LLC 3P Cyanide (free), 500 mL
Catalog No. NC0292660
Encompass_Preferred
(NSI by Zeptometrix) 1000mg/L in Dilute NaOH. Derived from high purity Potassium cyanide, NIST traceable.
Catalog No. NC0292660 Supplier SPEX CERTIPREP LLC 3P Supplier No. IS029500
(NSI by Zeptometrix) 1000mg/L in Dilute NaOH. Derived from high purity Potassium cyanide, NIST traceable.