Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanidin-3-O-arabinoside | 111613-04-8 | 95.7% | 1 MG
SDP

Catalog No. 5000339369
Encompass_Preferred

Cyanidin-3-O-arabinoside is an antioxidant agent and a pigment isolated from flowers of *Rhododendron cv. Lems Stormcloud*. It is for research use only.

  • Antioxidant agent
  • Pigment isolated from flowers of Rhododendron cv. Lems Stormcloud
  • For research use only
  • Purity of 95.71%
  • Solid appearance
  • Brown to black color
  • Store at -20°C in sealed container, away from moisture and light
  • In solvent, store at -80°C for 6 months, or -20°C for 1 month in sealed container, away from moisture and light

Catalog No. 50-003-39369 Supplier Medchemexpress LLC Supplier No. HY1291391MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanidin-3-O-arabinoside is an antioxidant agent and a pigment isolated from flowers of *Rhododendron cv. Lems Stormcloud*. It is for research use only.

  • Antioxidant agent
  • Pigment isolated from flowers of Rhododendron cv. Lems Stormcloud
  • For research use only
  • Purity of 95.71%
  • Solid appearance
  • Brown to black color
  • Store at -20°C in sealed container, away from moisture and light
  • In solvent, store at -80°C for 6 months, or -20°C for 1 month in sealed container, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.