Learn More
Medchemexpress LLC Cyanidin-3-O-arabinoside | 111613-04-8 | MFCD27952825 | 95.7% | 454.81 g/mol | C20H19ClO10 | 5 MG
SDP
Catalog No. 5000263120
Encompass_Preferred
Cyanidin-3-O-arabinoside is an anthocyanin pigment isolated from the flowers of Rhododendron cv. Lems Stormcloud. Supplied as a research-grade material, the compound is provided as a 5 mg sample with manufacturer-reported purity and supporting documentation for laboratory use.
  • Anthocyanin pigment with antioxidant activity.
  • Molecular formula C20H19ClO10 and molecular weight 454.81 g/mol.
  • Supplied as a 5 mg sample with reported purity 95.7%.
  • Manufacturer datasheet, COA, and SDS available for quality and handling information.
  • Intended for biochemical assays and antioxidant research applications.
Catalog No. 50-002-63120 Supplier Medchemexpress LLC Supplier No. HY1291395MG
Cyanidin-3-O-arabinoside is an anthocyanin pigment isolated from the flowers of Rhododendron cv. Lems Stormcloud. Supplied as a research-grade material, the compound is provided as a 5 mg sample with manufacturer-reported purity and supporting documentation for laboratory use.
  • Anthocyanin pigment with antioxidant activity.
  • Molecular formula C20H19ClO10 and molecular weight 454.81 g/mol.
  • Supplied as a 5 mg sample with reported purity 95.7%.
  • Manufacturer datasheet, COA, and SDS available for quality and handling information.
  • Intended for biochemical assays and antioxidant research applications.