Learn More
TARGETMOL CHEMICALS INC Cyanidin-3-O-galactoside chlor
SDP
Catalog No. 502875837
Encompass_Preferred
Also available in 1 mL, 1 mg and bulk. Please contact Fisher for quotes. Cyanidin-3-O-galactoside chloride (Ideain chloride), extracted from hawthorn pericarp (EPHF), possesses antioxidant activity and is applicable in obesity research. Purity 99.87%
Catalog No. 50-287-5837 Supplier TARGETMOL CHEMICALS INC Supplier No. TN11065MG
Also available in 1 mL, 1 mg and bulk. Please contact Fisher for quotes. Cyanidin-3-O-galactoside chloride (Ideain chloride), extracted from hawthorn pericarp (EPHF), possesses antioxidant activity and is applicable in obesity research. Purity 99.87%