Learn More
Medchemexpress LLC Cyanidin-3-O-galactoside chloride | 27661-36-5 | 5 MG
SDP
Catalog No. 5000417791
Encompass_Preferred
Cyanidin-3-O-galactoside chloride is a component derived from the extract peel of hawthorn fruit (EPHF), demonstrating potent Acetylcholinesterase (AChE) inhibitory activity.
  • Exhibits strong antioxidant activity
  • Shows potent acetylcholinesterase (AChE) inhibitory activity
  • Displays high cytotoxicity against human tumor cell lines MCF-7 and SKOV-3
Catalog No. 50-004-17791 Supplier Medchemexpress LLC Supplier No. HYN41425MG
Cyanidin-3-O-galactoside chloride is a component derived from the extract peel of hawthorn fruit (EPHF), demonstrating potent Acetylcholinesterase (AChE) inhibitory activity.
  • Exhibits strong antioxidant activity
  • Shows potent acetylcholinesterase (AChE) inhibitory activity
  • Displays high cytotoxicity against human tumor cell lines MCF-7 and SKOV-3