Learn More
Medchemexpress LLC Cyanidin-3-O-galactoside chloride | 27661-36-5 | 99.2% | 484.84 | 1 MG
SDP
Catalog No. 5000365485
Encompass_Preferred
Cyanidin-3-O-galactoside chloride, also known as Ideain chloride, is a component found in the extract peel of hawthorn fruit (EPHF). EPHF exhibits strong Acetylcholinesterase (AChE) inhibitory activity, and also demonstrates significant antioxidant properties. It shows cytotoxicity against MCF-7 and SKOV-3 human tumor cell lines in a dose-dependent manner.
  • Exhibits strong AChE inhibitory activity
  • Demonstrates strong antioxidant activity
  • Shows high cytotoxicity against MCF-7 and SKOV-3 human tumor cell lines
Catalog No. 50-003-65485 Supplier Medchemexpress LLC Supplier No. 1MGHYN4142
Cyanidin-3-O-galactoside chloride, also known as Ideain chloride, is a component found in the extract peel of hawthorn fruit (EPHF). EPHF exhibits strong Acetylcholinesterase (AChE) inhibitory activity, and also demonstrates significant antioxidant properties. It shows cytotoxicity against MCF-7 and SKOV-3 human tumor cell lines in a dose-dependent manner.
  • Exhibits strong AChE inhibitory activity
  • Demonstrates strong antioxidant activity
  • Shows high cytotoxicity against MCF-7 and SKOV-3 human tumor cell lines