Learn More
Apexbio Technology LLC Cyanidin-3-O-glucoside chloride(Synonyms: Cyanidin-3-glucoside chloride, Chrysanthemin, Kuromanin chloride, Cyanidin 3-O-beta-D-glucopyranoside chloride), 20mg, CAS: 7084-24-4.
SDP
Catalog No. 5000061312
Encompass_Preferred
Cyanidin-3-O-glucoside chloride (CAS 7084-24-4) is an anthocyanin compound isolated from the pericarp of Glycine max (L ) Merr Structurally it belongs to flavonoid derivatives exhibiting antioxidative and anti-inflammatory properties through modulation of cellular signaling pathways such as NF- B and MAPK Research demonstrates its bioactivity in vitro and in vivo including the attenuation of oxidative stress suppression of inflammatory cytokines and beneficial effects on metabolic regulation It is utilized extensively in biomedical research to investigate oxidative and inflammatory mechanisms underlying chronic diseases Store in tightly sealed cool and dry conditions to maintain stability
Catalog No. 50-000-61312 Supplier Apexbio Technology LLC Supplier No. N256220
Cyanidin-3-O-glucoside chloride (CAS 7084-24-4) is an anthocyanin compound isolated from the pericarp of Glycine max (L ) Merr Structurally it belongs to flavonoid derivatives exhibiting antioxidative and anti-inflammatory properties through modulation of cellular signaling pathways such as NF- B and MAPK Research demonstrates its bioactivity in vitro and in vivo including the attenuation of oxidative stress suppression of inflammatory cytokines and beneficial effects on metabolic regulation It is utilized extensively in biomedical research to investigate oxidative and inflammatory mechanisms underlying chronic diseases Store in tightly sealed cool and dry conditions to maintain stability