Learn More
Medchemexpress LLC Cyanidin Chloride | 528-58-5 | 322.70 | 1 MG
SDP
Catalog No. 5000389350
Encompass_Preferred
Cyanidin Chloride, a subclass of anthocyanin, displays antioxidant and anti-carcinogenesis properties. It inhibits osteoclast formation, hydroxyapatite resorption, and receptor activator of NF-κB ligand (RANKL)-induced osteoclast marker gene expression.
  • Displays antioxidant properties.
  • Shows anti-carcinogenesis properties.
  • Inhibits osteoclast formation.
  • Inhibits hydroxyapatite resorption.
  • Inhibits receptor activator of NF-κB ligand (RANKL)-induced osteoclast marker gene expression.
  • Protects against ovariectomy-induced bone loss in an osteoporosis mouse model.
Catalog No. 50-003-89350 Supplier Medchemexpress LLC Supplier No. HYN04991MG
Cyanidin Chloride, a subclass of anthocyanin, displays antioxidant and anti-carcinogenesis properties. It inhibits osteoclast formation, hydroxyapatite resorption, and receptor activator of NF-κB ligand (RANKL)-induced osteoclast marker gene expression.
  • Displays antioxidant properties.
  • Shows anti-carcinogenesis properties.
  • Inhibits osteoclast formation.
  • Inhibits hydroxyapatite resorption.
  • Inhibits receptor activator of NF-κB ligand (RANKL)-induced osteoclast marker gene expression.
  • Protects against ovariectomy-induced bone loss in an osteoporosis mouse model.