Learn More
Aat Bioquest Cyanine 3.5 acid [equivalent to Cy3.5 acid]
SDP
Catalog No. 502979187
Encompass_Preferred
A variety of cyanine 3.5 (Cy3.5) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
Catalog No. 50-297-9187 Supplier Aat Bioquest Supplier No. 147
A variety of cyanine 3.5 (Cy3.5) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.