Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanine 3.5 bromide | 98.6% | 734.72 g·mol⁻1 | C42H44BrN3O4 | 10 MG
SDP

Catalog No. 5000297187
Encompass_Preferred

Cyanine 3.5 bromide is a red reactive fluorescent dye used for covalent labeling of primary amines on peptides, proteins, and oligonucleotides for fluorescence imaging and detection. It exhibits red fluorescence with excitation near 591 nm and emission near 604 nm, and is typically supplied as a small research-scale powdered quantity.

  • Reactive NHS ester for covalent labeling of primary amines.
  • Red fluorescence with typical excitation ~591 nm and emission ~604 nm.
  • Molecular formula C42H44BrN3O4, molecular weight 734.72 g·mol-1.
  • Purity reported around 98.6%.
  • Supplied as a 10 mg powder; store at -20 °C and protect from light.
  • Suitable for labeling peptides, proteins, and oligonucleotides in imaging applications.
  • Stable in common organic solvents; handle with appropriate safety precautions.

Catalog No. 50-002-97187 Supplier Medchemexpress LLC Supplier No. HYD1596A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanine 3.5 bromide is a red reactive fluorescent dye used for covalent labeling of primary amines on peptides, proteins, and oligonucleotides for fluorescence imaging and detection. It exhibits red fluorescence with excitation near 591 nm and emission near 604 nm, and is typically supplied as a small research-scale powdered quantity.

  • Reactive NHS ester for covalent labeling of primary amines.
  • Red fluorescence with typical excitation ~591 nm and emission ~604 nm.
  • Molecular formula C42H44BrN3O4, molecular weight 734.72 g·mol-1.
  • Purity reported around 98.6%.
  • Supplied as a 10 mg powder; store at -20 °C and protect from light.
  • Suitable for labeling peptides, proteins, and oligonucleotides in imaging applications.
  • Stable in common organic solvents; handle with appropriate safety precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.