Learn More
Apexbio Technology LLC Cyanine 3-AA-CTP(Synonyms: Cy3-AA-CTP, Cy3-labeled CTP, Cyanine 3-labeled cytidine triphosphate), 100ul (10 mM).
SDP
Catalog No. 5000060398
Encompass_Preferred
Cyanine 3-AA-CTP is a fluorescently labeled cytidine triphosphate derivative conjugated with the fluorophore Cy3 During in vitro transcription reactions Cyanine 3-AA-CTP is enzymatically incorporated into RNA strands producing Cy3-labeled RNA molecules suitable for fluorescence-based analyses Due to Cy3 s high fluorescence intensity and photostability the labeledRNAs facilitate diverse studies such as fluorescence microscopy imaging RNA-protein interaction assays and sensitive RNA detection methodologies Consequently Cyanine 3-AA-CTP serves as an important reagent for investigating RNA biology and function
Catalog No. 50-000-60398 Supplier Apexbio Technology LLC Supplier No. B81630.1
Cyanine 3-AA-CTP is a fluorescently labeled cytidine triphosphate derivative conjugated with the fluorophore Cy3 During in vitro transcription reactions Cyanine 3-AA-CTP is enzymatically incorporated into RNA strands producing Cy3-labeled RNA molecules suitable for fluorescence-based analyses Due to Cy3 s high fluorescence intensity and photostability the labeledRNAs facilitate diverse studies such as fluorescence microscopy imaging RNA-protein interaction assays and sensitive RNA detection methodologies Consequently Cyanine 3-AA-CTP serves as an important reagent for investigating RNA biology and function