Learn More
Aat Bioquest Cyanine 3 amine [equivalent to Cy3 amine]
SDP
Catalog No. 502979122
Encompass_Preferred
A variety of cyanine 3 (Cy3) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
Catalog No. 50-297-9122 Supplier Aat Bioquest Supplier No. 145
A variety of cyanine 3 (Cy3) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.