Learn More
Enzo Life Sciences Cyanine 3 dUTP (25 nmol)
SDP
Catalog No. 501031806
Encompass_Preferred
Alternative Name: Cyanine 3-deoxyuridine-5'-triphosphate. Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Dark pink liquid. Use/Stability: Stable for at least one year after receipt when stored as recommended. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C Excitation maximum: 550 nm Emission maximum: 564 nm. Handling: Protect from light. Avoid freeze/thaw cycles.
Catalog No. 50-103-1806 Supplier Enzo Life Sciences Supplier No. ENZ42501
Alternative Name: Cyanine 3-deoxyuridine-5'-triphosphate. Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Dark pink liquid. Use/Stability: Stable for at least one year after receipt when stored as recommended. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C Excitation maximum: 550 nm Emission maximum: 564 nm. Handling: Protect from light. Avoid freeze/thaw cycles.