Learn More
Aat Bioquest Cyanine 3 monosuccinimidyl ester [equivalent to Cy3® NHS ester]
SDP
Supplier:  Aat Bioquest 141
Encompass_Preferred
A variety of cyanine 3 (Cy3®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. They are widely used for labeling peptides, proteins and oligos etc.
Catalog No. 50-195-2751