Learn More
Enzo Life Sciences Cyanine 3 UTP enhanced (250 nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42505
Encompass_Preferred
Cyanine 3-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP . Alternative Name: Cyanine 3-uridine-5'-triphosphate (enhanced). Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Dark pink liquid. Use/Stability: Stable for at least one year after receipt when stored as recommended. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C Excitation maximum: 550 nm Emission maximum: 564 nm. Handling: Protect from light. Avoid freeze/thaw cycles.
Catalog No. 50-103-1808