Learn More
Aat Bioquest Cyanine 5.5 hydrazide [equivalent to Cy5.5® hydrazide]
SDP
Catalog No. 501952787
Encompass_Preferred
A variety of cyanine 5.5 (Cy5.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. They are widely used for labeling peptides, proteins and oligos etc.
Catalog No. 50-195-2787 Supplier Aat Bioquest Supplier No. 177
A variety of cyanine 5.5 (Cy5.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. They are widely used for labeling peptides, proteins and oligos etc.