Learn More
Aat Bioquest Cyanine 5.5 monoacid [equivalent to Cy5.5 acid]
SDP
Catalog No. 502979150
Encompass_Preferred
A variety of cyanine 5.5 (Cy5.5) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
Catalog No. 50-297-9150 Supplier Aat Bioquest Supplier No. 173
A variety of cyanine 5.5 (Cy5.5) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.