Learn More
Aat Bioquest Cyanine 5 alkyne [equivalent to Cy5 alkyne]
SDP
Catalog No. 502979188
Encompass_Preferred
A variety of cyanine 5 ( Cy5) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
Catalog No. 50-297-9188 Supplier Aat Bioquest Supplier No. 154
A variety of cyanine 5 ( Cy5) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.