Learn More
Enzo Life Sciences Cyanine 5 dUTP (25 nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42502
Encompass_Preferred
Alternative Name: Cyanine 5-deoxyuridine-5'-triphosphate. Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Blue liquid. Use/Stability: Stable for at least one year after receipt when stored as recommended. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C Excitation maximum: 650 nm Emission maximum: 662 nm. Handling: Protect from light. Avoid freeze/thaw cycles.
Catalog No. 50-103-1807