Learn More
Biotium Cyanine 555 Tyramide
Catalog No. 501964998
Encompass_Preferred
Cyanine 555 tyramide (Ex/Em 555/565 nm) can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. Cyanine 555 is structurally identical to Cy3 from GE Healthcare.
Catalog No. 50-196-4998 Supplier Biotium Supplier No. 96020
Cyanine 555 tyramide (Ex/Em 555/565 nm) can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. Cyanine 555 is structurally identical to Cy3 from GE Healthcare.