Learn More
Biotium Cyanine 647 Succinimidyl Ester
Catalog No. 501964806
Encompass_Preferred
Biotium offers high quality amine-reactive succinimidyl ester (SE, or NHS-Ester) dyes used to label proteins, oligonucleotides, and nucleic acids. We offer Cyanine 555 (Cy3), Cyanine 647 (Cy5), Cyanine 770 (Cy5.5), and Cyanine 750 (Cy7) as high quality succinimidyl esters. Also see our CF Dye Succinimidyl Ester, available with more than 20 bright and photostable CF Dyes. Cyanine 647 is structurally identical to Cy5 from GE Healthcare. Properties: Ex / Em (water) = 650/665 nm; Dark blue solid soluble in DMF and DMSO; Store at -20°C and protect from moisture and light; C 43 H 58 N 4 O 10 S 2; MW: 855. Cy3, Cy5, Cy5.5, and Cy7 are registered trademarks of GE Healthcare.
Catalog No. 50-196-4806 Supplier Biotium Supplier No. 900251MG
Biotium offers high quality amine-reactive succinimidyl ester (SE, or NHS-Ester) dyes used to label proteins, oligonucleotides, and nucleic acids. We offer Cyanine 555 (Cy3), Cyanine 647 (Cy5), Cyanine 770 (Cy5.5), and Cyanine 750 (Cy7) as high quality succinimidyl esters. Also see our CF Dye Succinimidyl Ester, available with more than 20 bright and photostable CF Dyes. Cyanine 647 is structurally identical to Cy5 from GE Healthcare. Properties: Ex / Em (water) = 650/665 nm; Dark blue solid soluble in DMF and DMSO; Store at -20°C and protect from moisture and light; C 43 H 58 N 4 O 10 S 2; MW: 855. Cy3, Cy5, Cy5.5, and Cy7 are registered trademarks of GE Healthcare.