Learn More
Aat Bioquest Cyanine 7 monoacid [equivalent to Cy7 acid]
SDP
Catalog No. 502979214
Encompass_Preferred
A variety of cyanine 7 (Cy7) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
Catalog No. 50-297-9214 Supplier Aat Bioquest Supplier No. 160
A variety of cyanine 7 (Cy7) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.