Learn More
Medchemexpress LLC Cyanine2 | 260430-02-2 | 98.0% | 418.49 g/mol | C25H26N2O4 | 10 MG
SDP
Catalog No. 5000298025
Encompass_Preferred
Cy2 is a cyanine-class fluorescent dye for labeling biomolecules and fluorescence imaging. It is excited at 488 nm and emits at 515 nm, with high purity and good solubility in DMSO; store protected from light at -20°C as a solid, or in solution at -80°C for long-term storage.
Catalog No. 50-002-98025 Supplier Medchemexpress LLC Supplier No. HYD082710MG
Cy2 is a cyanine-class fluorescent dye for labeling biomolecules and fluorescence imaging. It is excited at 488 nm and emits at 515 nm, with high purity and good solubility in DMSO; store protected from light at -20°C as a solid, or in solution at -80°C for long-term storage.