Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanine2 (iodine) | 186205-37-8 | 96.8% | 546.40 g·mol⁻¹ | C25H27IN2O4 | 10 MG
SDP

Catalog No. 5000297350
Encompass_Preferred

Cy2 (iodine) is a cyanine-class fluorescent dye used for labeling proteins, antibodies, and small molecules. It provides long-wavelength absorption and emission, a high extinction coefficient, and good water solubility, enabling efficient conjugation and fluorescence imaging applications.

  • High purity suitable for research use.
  • Molecular weight 546.40 g·mol⁻¹.
  • Chemical formula C25H27IN2O4.
  • Available in small milligram packages for conjugation workflows.
  • Analytical documentation available: COA, SDS, HNMR, RP-HPLC, and MS.
  • Typical application: labeling of proteins and antibodies for fluorescence assays.

Catalog No. 50-002-97350 Supplier Medchemexpress LLC Supplier No. HYD105410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cy2 (iodine) is a cyanine-class fluorescent dye used for labeling proteins, antibodies, and small molecules. It provides long-wavelength absorption and emission, a high extinction coefficient, and good water solubility, enabling efficient conjugation and fluorescence imaging applications.

  • High purity suitable for research use.
  • Molecular weight 546.40 g·mol⁻¹.
  • Chemical formula C25H27IN2O4.
  • Available in small milligram packages for conjugation workflows.
  • Analytical documentation available: COA, SDS, HNMR, RP-HPLC, and MS.
  • Typical application: labeling of proteins and antibodies for fluorescence assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.