Learn More
Medchemexpress LLC Cyanine3 DBCO hexafluorophosphate | 2692677-79-3 | 95.0% | 902.99 | C51H57F6N4O2P | 10 MG
SDP
Catalog No. 5000298846
Encompass_Preferred
Cyanine3 DBCO hexafluorophosphate is an azide-reactive fluorescent probe designed for copper-free click chemistry labeling and imaging of azide-tagged biomolecules. The DBCO moiety enables strain-promoted azide-alkyne cycloaddition without a copper catalyst, while the Cyanine3 dye provides bright visible fluorescence for microscopy and detection applications.
  • Enables copper-free click chemistry labeling of azide-tagged biomolecules.
  • Provides bright Cy3 fluorescence for imaging and detection.
  • Supplied as a hexafluorophosphate salt at high purity for reliable performance.
  • Stable when stored protected from light at -20°C; in solvent, store at -80°C for long-term use.
  • Suitable for biomolecule labeling, microscopy, and fluorescence-based assays.
Catalog No. 50-002-98846 Supplier Medchemexpress LLC Supplier No. HYD162210MG
Cyanine3 DBCO hexafluorophosphate is an azide-reactive fluorescent probe designed for copper-free click chemistry labeling and imaging of azide-tagged biomolecules. The DBCO moiety enables strain-promoted azide-alkyne cycloaddition without a copper catalyst, while the Cyanine3 dye provides bright visible fluorescence for microscopy and detection applications.
  • Enables copper-free click chemistry labeling of azide-tagged biomolecules.
  • Provides bright Cy3 fluorescence for imaging and detection.
  • Supplied as a hexafluorophosphate salt at high purity for reliable performance.
  • Stable when stored protected from light at -20°C; in solvent, store at -80°C for long-term use.
  • Suitable for biomolecule labeling, microscopy, and fluorescence-based assays.