Learn More
Medchemexpress LLC Cyanine5.5 amine | 2097714-45-7 | 753.88 | C46H58Cl2N4O | 10 MG
SDP
Catalog No. 5000297186
Encompass_Preferred
Cyanine5.5 amine is a near-infrared fluorescent dye bearing a reactive amine group for conjugation to biomolecules, nanoparticles, and polymers. It exhibits excitation at 680 nm and emission at 710 nm, making it suitable for low-background imaging in confocal and fluorescence microscopy applications.
  • Near-infrared excitation and emission (680 nm/710 nm) for reduced autofluorescence.
  • Amine functional group enables straightforward conjugation to proteins and nanoparticles.
  • Appropriate for confocal and fluorescence microscopy and fluorescence-based tracking.
  • Available in small and bulk pack sizes to support scouting experiments and scale-up.
Catalog No. 50-002-97186 Supplier Medchemexpress LLC Supplier No. HYD154010MG
Cyanine5.5 amine is a near-infrared fluorescent dye bearing a reactive amine group for conjugation to biomolecules, nanoparticles, and polymers. It exhibits excitation at 680 nm and emission at 710 nm, making it suitable for low-background imaging in confocal and fluorescence microscopy applications.
  • Near-infrared excitation and emission (680 nm/710 nm) for reduced autofluorescence.
  • Amine functional group enables straightforward conjugation to proteins and nanoparticles.
  • Appropriate for confocal and fluorescence microscopy and fluorescence-based tracking.
  • Available in small and bulk pack sizes to support scouting experiments and scale-up.