Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanine5.5 amine | 2097714-45-7 | 99.3% | 753.88 | 100 MG
SDP

Catalog No. 5000431240
Encompass_Preferred

Cyanine5.5 amine is a Cy5.5 analogue and a near-infrared (NIR) fluorescent dye (Ex=648 nm, Em=710 nm). It can be used in the preparation of Cy5.5-labeled nanoparticles for tracking and imaging with low fluorescence background using confocal microscopy.

  • Solid appearance
  • Purple to purplish red color
  • Excitation wavelength: 648 nm
  • Emission wavelength: 710 nm
  • Soluble in DMSO at 100 mg/mL
  • Store at -20°C, sealed, away from moisture and light
  • Suitable for research use only

Catalog No. 50-004-31240 Supplier Medchemexpress LLC Supplier No. HYD1540100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanine5.5 amine is a Cy5.5 analogue and a near-infrared (NIR) fluorescent dye (Ex=648 nm, Em=710 nm). It can be used in the preparation of Cy5.5-labeled nanoparticles for tracking and imaging with low fluorescence background using confocal microscopy.

  • Solid appearance
  • Purple to purplish red color
  • Excitation wavelength: 648 nm
  • Emission wavelength: 710 nm
  • Soluble in DMSO at 100 mg/mL
  • Store at -20°C, sealed, away from moisture and light
  • Suitable for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.