Learn More
Medchemexpress LLC Cyanine5.5 NHS ester chloride | 1469277-96-0 | MFCD28385524 | 95.7% | 716.31 g·mol-1 | C44H46ClN3O4 | 5 MG
SDP
Catalog No. 5000222334
Encompass_Preferred
Cyanine5.5 NHS ester chloride is a near-infrared reactive dye supplied as a solid NHS-activated ester that covalently labels primary amines in proteins, peptides, antibodies, and small molecules for fluorescence detection and imaging. The reagent is provided with analytical documentation to support purity and identity.
Catalog No. 50-002-22334 Supplier Medchemexpress LLC Supplier No. HY1354155MG
Cyanine5.5 NHS ester chloride is a near-infrared reactive dye supplied as a solid NHS-activated ester that covalently labels primary amines in proteins, peptides, antibodies, and small molecules for fluorescence detection and imaging. The reagent is provided with analytical documentation to support purity and identity.