Learn More
Medchemexpress LLC Cyanine5.5 NHS ester (tetrafluoroborate) | 2375105-86-3 | 98.4% | 767.66 | 10 MG
SDP
Catalog No. 5000365466
Encompass_Preferred
Cyanine5.5 NHS ester tetrafluoroborate is a member of the cyanine dye series, serving as a common fluorescent marker for biomolecules. It can interact with biomolecules and may also bind to double-helical DNA through intercalation, exhibiting enhanced fluorescence upon binding.
  • Common fluorescent marker for biomolecules
  • Interacts with biomolecules
  • May bind to double-helical DNA through intercalation
  • Exhibits enhanced fluorescence upon binding to DNA
  • For research use only
  • In vitro protocols available for protein preparation, dye preparation, dosage calculation, conjugation reactions, and purification
Catalog No. 50-003-65466 Supplier Medchemexpress LLC Supplier No. 10MGHYD1322
Cyanine5.5 NHS ester tetrafluoroborate is a member of the cyanine dye series, serving as a common fluorescent marker for biomolecules. It can interact with biomolecules and may also bind to double-helical DNA through intercalation, exhibiting enhanced fluorescence upon binding.
  • Common fluorescent marker for biomolecules
  • Interacts with biomolecules
  • May bind to double-helical DNA through intercalation
  • Exhibits enhanced fluorescence upon binding to DNA
  • For research use only
  • In vitro protocols available for protein preparation, dye preparation, dosage calculation, conjugation reactions, and purification